Advantec GA20047MM Grade GA200 Glass Fiber Filters; 47mm dia; 50/box

Representative image only.

Advantec GA20047MM Grade GA200 Glass Fiber Filters; 47mm dia; 50/box

Advantec | Mfr: GA20047MM| Item: 0665762

For analytical or gravimetric studies

₹9,000.00INR / Pkg of 50

₹9,000.00INR / Pkg of 50
Product Description

Key Features

  • 1 year warranty

More About this Item

Thick Filter; filtering viscous fluids such as liquid sugars and gels.

1 Year Warranty
Specifications
  • MaterialGlass Microfiber
  • Diameter (mm)47
  • Diameter (cm)4.7
  • DescriptionGrade GA200 Glass Fiber Filters; 47mm dia; 50/box
  • Warranty1 year
Reviews
Q&A