Advantec DP7025MM Grade DP70 Glass Fiber Filters; 25mm dia; 50/box

Representative image only.

Advantec DP7025MM Grade DP70 Glass Fiber Filters; 25mm dia; 50/box

Advantec | Mfr: DP7025MM| Item: 0665800

For analytical or gravimetric studies

₹4,100.00INR / Pkg of 50

₹4,100.00INR / Pkg of 50
Product Description

Key Features

  • 1 year warranty

More About this Item

High wet strength and very high loading capacity; dust and fine particulate sampling.

1 Year Warranty
Specifications
  • MaterialGlass Microfiber
  • Diameter (mm)25
  • Diameter (cm)2.5
  • DescriptionGrade DP70 Glass Fiber Filters; 25mm dia; 50/box
  • Warranty1 year
Reviews
Q&A