Advantec 800000 Forceps, 304 Stainless Steel
Representative image only.
Advantec 800000 Forceps, 304 Stainless Steel
Advantec | Mfr: 17800000| Item: 0290250
Make membrane handling easier with forceps
₹10,700.00INR / Each
₹10,700.00INR / Each
Product Description
Key Features
  • 304 stainless steel construction
  • Beveled tips and unserrated
  • Nonslip handle provides a secure grip
  • Sterilize ends vis flame
  • 1 year warranty
More About this Item
These 304 SS forceps are designed for precise handling of filters and membranes in laboratory applications. Ergonomic nonslip handle design offers secure and comfortable handling.
1 Year Warranty
Specifications
  • Material304 Stainless Steel
  • Compatible WithFilters and membranes
  • Description304 Stainless Steel Forceps