Advantec 357210 Silicone O-Ring for SS Filter Holders, 47 mm
Representative image only.
Advantec 357210 Silicone O-Ring for SS Filter Holders, 47 mm
Advantec | Mfr: 357210| Item: 0293060
Ensure a tight fit with O-rings
₹3,200.00INR / Each
₹3,200.00INR / Each
Product Description
Key Features
  • Silicone construction
  • 1 year warranty
More About this Item
Use with stainless steel in-line filter holder 02929-20.
1 Year Warranty
Specifications
  • MaterialSilicone
  • Compatible WithStainless steel in-line filter holder 02929-20
  • DescriptionSilicone O-Ring for SS Filter Holders, 47 mm