Advantec 341012 Vent Relief Valve and Washer for Stirred Cell Device, UHP
Representative image only.
Advantec 341012 Vent Relief Valve and Washer for Stirred Cell Device, UHP
Advantec | Mfr: 341012| Item: 0291046
Vents to relieve pressure
₹248,000.00INR / Each
₹248,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Replacement pressure relief valve for 02910-series Advantec stirred cells.
1 Year Warranty
Specifications
  • Compatible With02910-series stirred cells
  • DescriptionVent Relief Valve and Washer for Stirred Cell Device, UHP