Advantec 311425 Receiver Flask for All-Glass Filtration Assemblies, 1L Erlenmeyer

Representative image only.

Advantec 311425 Receiver Flask for All-Glass Filtration Assemblies, 1L Erlenmeyer

Advantec | Mfr: 311425| Item: 0664551

All wetted surfaces are pure borosilicate glass

₹111,000.00INR / Each

₹111,000.00INR / Each
Product Description

Key Features

  • Constructed of borosilicate glass
  • Ground glass jount 40/35 with 6 mm side arm
  • 1 year warranty

More About this Item

This replacement 1000 mL receiver flask is compatible with Advantecx All-Glass Filtration Assemblies 06645-49, -65, and -41.

1 Year Warranty
Specifications
  • DescriptionReceiver Flask for All-Glass Filtration Assemblies, 1L Erlenmeyer
  • Warranty1 year
Reviews
Q&A