Advantec 304702 PTFE Gasket for SS Filter Holders, LS47
Representative image only.
Advantec 304702 PTFE Gasket for SS Filter Holders, LS47
Advantec | Mfr: 304702| Item: 0293040
Ensure a tight fit with O-rings
₹4,700.00INR / Each
₹4,700.00INR / Each
Product Description
Key Features
  • PTFE construction
  • 1 year warranty
More About this Item
Use with stainless steel in-line filter holder 02929-20.
1 Year Warranty
Specifications
  • MaterialPTFE
  • Compatible WithStainless steel in-line filter holder 02929-20
  • DescriptionPTFE Gasket for SS Filter Holders, LS47