Advantec 07030041 Test Strips, Chrome Check (Cr6+), 0 to 200 ppm; 25 Strips/Pk

Representative image only.

Advantec 07030041 Test Strips, Chrome Check (Cr6+), 0 to 200 ppm; 25 Strips/Pk

Advantec | Mfr: 07030041| Item: 5920116

Color-change test papers give rapid indication of pH level ion presence

₹4,400.00INR / Pkg of 25

₹4,400.00INR / Pkg of 25

Buy It Together

Current Item

Advantec 07030041 Test Strips, Chrome Check (Cr6+), 0 to 200 ppm; 25 Strips/Pk
Advantec 07030041 Test Strips, Chrome Check (Cr6+), 0 to 200 ppm; 25 Strips/Pk
5920116

₹4,400.00INR / Pkg of 25
+ +
Reagents Potassium Iodide, 25% (w/v), 1L, Volumetric
7891805

₹24,900.00INR / Each
+
Reagents Conductivity Standard 1413 µS/cm, 1L
7891521

₹11,400.00INR / Each
+ +
LabChem Nitric Acid, ACS Grade; 1 L
8002952

Call For Price
+
Environmental Express Oakton pH Indicator Strips, 0 to 14, 100/pk
3585002

₹5,300.00INR / Pkg of 100
+
LabChem Potassium Iodide, 10% (w/v); 500 mL
8003513

Call For Price
+
Reagents Hydrochloric Acid, 50% (v/v), 500mL, Volumetric
7891158

₹10,800.00INR / Each
+
LabChem Sodium Hydroxide, 10% (w/w); 1 L
8003839

Call For Price
=
Total Price 56800.00 ADD 10 ITEMS TO CART
Product Description

Key Features

  • Obtain quick ion concentration measurements in the field or laboratory
  • Dip and compare color change — no calibration, electricity, or warm up needed

More About this Item

These ion test strips make water testing fast and easy. Conveniently packed in screw top bottles, the test papers are safe even in rugged field applications. Obtaining simultanious pH and ion concentration results couldn't be easier. Simply dip the test strip in your water sample, wait about 15 seconds for the water to penetrate the strip, and compare the color changes against the chart on the bottle's label.

Specifications
  • TestChrome
  • Measurement Points0 to 200 ppm
  • Reagent PlatformStrips
  • Tests Per Kit25
  • DescriptionTest Strips, Chrome Check (Cr6+), 0 to 200 ppm; 25 Strips/Pk
Reviews
Q&A