Advantec 07030010 Test Strips, Alumi Check (Aluminum Al3+), 0 to 100 ppm; 100 Strips/Pk

Representative image only.

Advantec 07030010 Test Strips, Alumi Check (Aluminum Al3+), 0 to 100 ppm; 100 Strips/Pk

Advantec | Mfr: 07030010| Item: 5920110

Color-change test papers give rapid indication of pH level ion presence

₹18,100.00INR / Pkg of 100

₹18,100.00INR / Pkg of 100

Buy It Together

Current Item

Advantec 07030010 Test Strips, Alumi Check (Aluminum Al3+), 0 to 100 ppm; 100 Strips/Pk
Advantec 07030010 Test Strips, Alumi Check (Aluminum Al3+), 0 to 100 ppm; 100 Strips/Pk
5920110

₹18,100.00INR / Pkg of 100
+
Environmental Express Oakton pH Indicator Strips, 0 to 14, 100/pk
3585002

₹5,300.00INR / Pkg of 100
+
Reagents Hydrochloric Acid, 50% (v/v), 500mL, Volumetric
7891158

₹10,800.00INR / Each
+
Reagents Conductivity Standard 1413 µS/cm, 1L
7891521

₹11,400.00INR / Each
+
Reagents Potassium Iodide, 25% (w/v), 1L, Volumetric
7891805

₹24,900.00INR / Each
+
LabChem Nitric Acid, ACS Grade; 1 L
8002952

Call For Price
+
LabChem Potassium Iodide, 10% (w/v); 500 mL
8003513

Call For Price
+
LabChem Sodium Hydroxide, 10% (w/w); 1 L
8003839

Call For Price
+ + =
Total Price 70500.00 ADD 10 ITEMS TO CART
Product Description

Key Features

  • Obtain quick ion concentration measurements in the field or laboratory
  • Dip and compare color change — no calibration, electricity, or warm up needed

More About this Item

These ion test strips make water testing fast and easy. Conveniently packed in screw top bottles, the test papers are safe even in rugged field applications. Obtaining simultanious pH and ion concentration results couldn't be easier. Simply dip the test strip in your water sample, wait about 15 seconds for the water to penetrate the strip, and compare the color changes against the chart on the bottle's label.

Specifications
  • TestAluminum
  • Measurement Points0 to 100 ppm
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionTest Strips, Alumi Check (Aluminum Al3+), 0 to 100 ppm; 100 Strips/Pk
Reviews
Q&A