Advanced Scientific Sterile Single-Use Tank Liner, 264 gal.; 10/Pk
Representative image only.
Advanced Scientific Sterile Single-Use Tank Liner, 264 gal.; 10/Pk
Advanced Scientific | Mfr: B100053-I| Item: 0631014
Eliminate risk of cross contamination and save time
₹159,000.00INR / Pkg of 10
₹159,000.00INR / Pkg of 10
Buy It Together
Current Item
Advanced Scientific Sterile single-use tank liner, 264 gal.; 10/pk
Advanced Scientific Sterile single-use tank liner, 264 gal.; 10/pk
0631014
₹159,000.00INR / Pkg of 10
+
Chem-Tainer Low-Profile Cylindrical Tank, PE, 55 gallon
0692551
₹82,600.00INR / Each
=
Total Price 241600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • No more washing, cleaning, or revalidation of rigid, reusable tanks!
More About this Item
Reduce delays due to unavailability of clean tanks or drums. These sterile tank liners are ready for immediate use. Designed for the biopharmaceutical industry, these liners help eliminate risk of cross contamination. You save money by cutting labor costs and reducing the need for expensive equipment or dedicated tanks and drums. Liners are pyrogen free!
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • MaterialLLDPE
  • Volume (Gal)264
  • Volume (Liters)1000
  • Height (in)49
  • Height (cm)124.46
  • Outside Diameter (OD) (in)42
  • DescriptionSterile single-use tank liner, 264 gal.; 10/pk