Adam Equipment DU Software - Data Capture Utility for Balances and Scales
Representative image only.
Adam Equipment DU Software - Data Capture Utility for Balances and Scales
Adam Equipment | Mfr: ADAMDU DATA COLLECTION SOFTWARE| Item: 1112096
₹28,200.00INR / Each
₹28,200.00INR / Each
Buy It Together
Current Item
Adam Equipment DU Software - Data Capture Utility for Balances and Scales
Adam Equipment DU Software - Data Capture Utility for Balances and Scales
1112096
₹28,200.00INR / Each
+ =
Total Price 28200.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Easily capture data from up to 8 different balances
More About this Item
ADAM DU (Data Utility) is an application that allows you to quickly and easily capture data from an ADAM Laboratory Balance or Weighing Scale and perform various functions on the collected readings.
ADAM DU can perform various tasks, such as graph the data, perform basic mathematical statistical analysis, export the readings to several common file formats. Also quickly export data to other applications (e.g. MS Excel, MS Word or the Windows Clipboard). ADAM DU also provides basic remote control of the balance/scale*.
ADAM DU can collect data from up to 8 different balances/scales simultaneously, each data collection session can be individually monitored, configured and customised to your requirements.
Adam DU can also speak the readings received. This is ideal if you want to stay informed of a scale's progress whilst completing other tasks or maybe you might be visually impaired.
Specifications
  • Accessory TypeSoftware
  • DescriptionDU Software - Data Capture Utility for Balances and Scales