Adam Equipment Disposable Aluminum Sample Pans for Balances; 250/PK
Representative image only.
Adam Equipment Disposable Aluminum Sample Pans for Balances; 250/PK
Adam Equipment | Mfr: ALUMINUM PANS| Item: 1170066
Ideal for moisture analysis testing
₹58,600.00INR / Pkg of 250
₹58,600.00INR / Pkg of 250
Buy It Together
Current Item
Adam Equipment Disposable Aluminum Sample Pans for Balances; 250/PK
Adam Equipment Disposable Aluminum Sample Pans for Balances; 250/PK
1170066
₹58,600.00INR / Pkg of 250
+ =
Total Price 667600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Fits neatly on PMB moisture anayzers pan holder
More About this Item
To keep items being weighed in place, the disposable sample pans are 10 mm deep by 100 mm dia.
Disposable, aluminum weighing pans. 250 per pack.
Specifications
  • Accessory TypeSample pan
  • MaterialAluminum
  • Depth (in)2/5
  • Diameter (in)4
  • DescriptionDisposable Aluminum Sample Pans for Balances; 250/PK