Accuform MSMK118VS Sign, Danger-No Smoking Or Open Flames/Sparks, 7x10", Adhesive Vinyl; 1/Pk
Representative image only.
Accuform MSMK118VS Sign, Danger-No Smoking Or Open Flames/Sparks, 7x10", Adhesive Vinyl; 1/Pk
Accuform | Mfr: MSMK118VS| Item: 5071023
₹3,500.00INR / Each
₹3,500.00INR / Each
Product Description
More About this Item
These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
DANGER (red): Indicates an imminently hazardous situation which, if not avoided, will result in death or serious injury.
Adhesive vinyl is a flexible 4-mil pressure-sensitive vinyl with adhesive back and peel-off liner. This material is ideal for low-cost, light-duty applications.
Specifications
  • HeaderDANGER
  • LanguagesEnglish
  • MaterialVinyl
  • Width (in)10
  • Height (in)7
  • DescriptionSign, Danger-No Smoking Or Open Flames/Sparks, 7x10", Adhesive Vinyl; 1/Pk