Accuform MLT405CTP Lockout/Tagout Tag, Danger-Do Not Operate, 5.8x3.1", Cardstock; 25/Pk
Representative image only.
Accuform MLT405CTP Lockout/Tagout Tag, Danger-Do Not Operate, 5.8x3.1", Cardstock; 25/Pk
Accuform | Mfr: MLT405CTP| Item: 4050300
₹6,700.00INR / Pkg of 25
₹6,700.00INR / Pkg of 25
Buy It Together
Current Item
Accuform MLT405CTP Lockout/Tagout Tag, Danger-Do Not Operate, 5.8x3.1", Cardstock; 25/Pk
Accuform MLT405CTP Lockout/Tagout Tag, Danger-Do Not Operate, 5.8x3.1", Cardstock; 25/Pk
4050300
₹6,700.00INR / Pkg of 25
+
Accuform TAC800 Tag Center Board 4-3/4" H x 3-5/8" W x 2" D
4050400
₹54,300.00INR / Each
=
Total Price 61000.00 ADD 2 ITEMS TO CART
Product Description
More About this Item
Tags comply with tagout requirements of the OSHA Lockout/Tagout standard 29 CFR 1910.147. Use tags to mark and identify the lockout situation to prevent the accidental or unexpected start up or release of stored energy when working on equipment and machinery. Tags feature top gabled corners and 3/8” (9.5-mm) hole. All tags are sold in packages of 25.
Compliance: OSHA 29 CFR 1910.147
Cardstock is an economical, 10-mil polyolefin-based material containing no wood-based pulp paper.
Specifications
  • HeaderDANGER
  • LanguagesEnglish
  • MaterialPolyolefin
  • DescriptionLockout/Tagout Tag, Danger-Do Not Operate, 5.8x3.1", Cardstock; 25/Pk