Accuform MLT401LTP Tag, Danger Do Not Operate Maintenance Department, 5 3/4" X 3 1/16" Hs-laminate
Representative image only.
Accuform MLT401LTP Tag, Danger Do Not Operate Maintenance Department, 5 3/4" X 3 1/16" Hs-laminate
Accuform | Mfr: MLT401LTP| Item: 4050314
₹25,600.00INR / Pkg of 25
₹25,600.00INR / Pkg of 25
Buy It Together
Current Item
Accuform MLT401LTP Tag, Danger Do Not Operate Maintenance Department, 5 3/4" X 3 1/16" Hs-laminate
Accuform MLT401LTP Tag, Danger Do Not Operate Maintenance Department, 5 3/4" X 3 1/16" Hs-laminate
4050314
₹25,600.00INR / Pkg of 25
+
Accuform TAC800 Tag Center Board 4-3/4" H x 3-5/8" W x 2" D
4050400
₹54,300.00INR / Each
=
Total Price 79900.00 ADD 2 ITEMS TO CART
Product Description
More About this Item
Tags comply with tagout requirements of the OSHA Lockout/Tagout standard 29 CFR 1910.147. Use tags to mark and identify the lockout situation to prevent the accidental or unexpected start up or release of stored energy when working on equipment and machinery. Tags feature top gabled corners and 3/8” (9.5-mm) hole. All tags are sold in packages of 25.
Compliance: OSHA 29 CFR 1910.147
Laminate is a durable poly-tag material for abusive, dirty, and greasy environments. The poly-tag is sealed on both sides by thick rigid vinyl.
Specifications
  • HeaderDANGER
  • LanguagesEnglish
  • MaterialLaminate
  • DescriptionTag, Danger Do Not Operate Maintenance Department, 5 3/4" X 3 1/16" Hs-laminate