Accuform LZH605EV5 GHS Label, Gas Cylinder Pictogram, 1" x 1", adhesive poly, 500/roll

Representative image only.

Accuform LZH605EV5 GHS Label, Gas Cylinder Pictogram, 1" x 1", adhesive poly, 500/roll

Accuform | Mfr: LZH605EV5| Item: 5071283

₹16,300.00INR / Roll of 500

₹16,300.00INR / Roll of 500
Product Description

More About this Item

GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Gas Cylinder pictogram indicates the following potential hazard: gases under pressure.

Specifications
  • HeaderGAS CYLINDER
  • LanguagesEnglish
  • MaterialAdhesive Poly
  • Width (in)1
  • Height (in)1
  • Quantity500/roll
  • DescriptionGHS Label, Gas Cylinder Pictogram, 1" x 1", adhesive poly, 500/roll
Reviews
Q&A