Accuform KCC630 Group View Slot Lockout Box, Yellow 12-3/4"W x 10-1/2"H x 3-3/8"D
Representative image only.
Accuform KCC630 Group View Slot Lockout Box, Yellow 12-3/4"W x 10-1/2"H x 3-3/8"D
Accuform | Mfr: KCC630| Item: 4050426
Easily view the keys that are included in a lock out situation - yet they stay locked out!
₹62,200.00INR / Each
₹62,200.00INR / Each
Product Description
Key Features
  • Box accommodates 16 hanging keys and 27 padlocks
  • Padlocks are placed through the clear cover and the box to prevent removal of the cover
  • A padlock on the box usually signifies that a worker is on the job
  • Padlock removed means that the worker has completed their portion of the job
  • No access to the keys until the last padlock is removed
  • The bottom center padlock is usually the last padlock to be removed for inspection of the work before lockout release
  • Top slot is included in the cover to drop in additional keys once lockout procedure has started
  • Includes two rear mounting holes for easy installation
Specifications
  • MaterialImpact resistant polycarbonate
  • Height (in)10.5
  • Width (InchShort)12 3/4
  • Depth (InchShort)3
  • DescriptionGroup View Slot Lockout Box, Yellow 12-3/4"W x 10-1/2"H x 3-3/8"D