A&D Weighing TB:164 Power Adapter, 220 V for A and D Hc-I Portable Counting Scale
Representative image only.
A&D Weighing TB:164 Power Adapter, 220 V for A and D Hc-I Portable Counting Scale
A&D Weighing | Mfr: TB:164| Item: 0103866
Provides continues power for your counting scale
₹20,700.00INR / Each
₹20,700.00INR / Each
Product Description
Key Features
  • 2 year warranty
More About this Item
Keep your A&D Weighing HC-i portable counting scales up and running with this power adapter.
This 220 VAC power adapter is ideal for A&D Engineering HC-i portable counting scales 11113-96; -97; -98; and -99
Two year warranty
Specifications
  • DescriptionPower Adapter, 220 V for A and D Hc-I Portable Counting Scale