A&D Weighing TB:137 Balance Accessory Prower Adapter, 220 V

Representative image only.

A&D Weighing TB:137 Balance Accessory Prower Adapter, 220 V

A&D Weighing | Mfr: TB:137| Item: 1111465

₹14,900.00INR / Each

₹14,900.00INR / Each

Buy It Together

Current Item

A&D Weighing TB:137 Balance Accessory Prower Adapter, 220 V
A&D Weighing TB:137 Balance Accessory Prower Adapter, 220 V
1111465

₹14,900.00INR / Each
+
Environmental Express Electrode Storage Solution; 500 mL
0065304

₹3,700.00INR / Pint of 1
+
Environmental Express Buffer Solution, pH 12.46; 500 mL
0065412

₹2,100.00INR / Each
+ + + + + + =
Total Price 474500.00 ADD 9 ITEMS TO CART
Product Description

Key Features

  • 1 year warranty

More About this Item

Choose the adapter to meet your power needs.

1 Year Warranty
Specifications
  • DescriptionBalance Accessory Prower Adapter, 220 V
  • Warranty1 Year
Reviews
Q&A