A&D Weighing GXA-17 Large Glass Breeze Break With Built-In Fanless Ionizer (Static Eliminator)
Representative image only.
A&D Weighing GXA-17 Large Glass Breeze Break With Built-In Fanless Ionizer (Static Eliminator)
A&D Weighing | Mfr: GXA-17| Item: 1110957
₹427,000.00INR / Each
₹427,000.00INR / Each
Buy It Together
Current Item
A&D Weighing GXA-17 Large Glass Breeze Break With Built-In Fanless Ionizer (Static Eliminator)
A&D Weighing GXA-17 Large Glass Breeze Break With Built-In Fanless Ionizer (Static Eliminator)
1110957
₹427,000.00INR / Each
+ =
Total Price 427000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Compatible with A&D Apollo GF and GX series balances
  • 1 year warranty
More About this Item
Improve weighing accuracy and control your samples.
The fanless ionizer can neutralize even extremely fine powders without disturbance. The glass panes disappear into the back when opening the door, requiring no extra space at the rear of the balance. Easily attachable/detachable.
1 Year Warranty
Specifications
  • DescriptionLarge Glass Breeze Break With Built-In Fanless Ionizer (Static Eliminator)