A&D Weighing GXA-13 Density Determination Kit (For The 1 Mg Models Only)
Representative image only.
A&D Weighing GXA-13 Density Determination Kit (For The 1 Mg Models Only)
A&D Weighing | Mfr: GXA-13| Item: 1110979
Use your balance to determine density with these convenient kits
₹176,000.00INR / Each
₹176,000.00INR / Each
Buy It Together
Current Item
A&D Weighing GXA-13 Density Determination Kit (For The 1 Mg Models Only)
A&D Weighing GXA-13 Density Determination Kit (For The 1 Mg Models Only)
1110979
₹176,000.00INR / Each
+
Cole-Parmer RS-232 to USB Cable
1010083
₹9,900.00INR / Each
=
Total Price 185900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Expand the capabilities of your balance with these convenient kits
  • Ensure the kit corresponds to your correct A&D Weighing balance
  • Easily measure the density of solid or liquid when it is combined with electronic balances
  • 1 year warranty
More About this Item
These density determination kits, when combined with one of the A&D Weighing electronic balances, allows the density of a solid and liquid to be measured easily. Balances sold separately.
Compatible with A&D balances GX-203A, GX-303A, GX-403A, GX-603A, GX-1003A, GX-1603A, GF-203A, GF-303A, GF-403A, GF-603A, GF-1003A, GF-1603A.
1 Year Warranty
Specifications
  • DescriptionDensity Determination Kit (For The 1 Mg Models Only)