A&D Weighing GXA-06 Analog Output Interface
Representative image only.
A&D Weighing GXA-06 Analog Output Interface
A&D Weighing | Mfr: GXA-06| Item: 1110960
Communicate with your A&D balance using compatible output and relay kits
₹66,200.00INR / Each
₹66,200.00INR / Each
Product Description
Key Features
  • Compatible with A&D GF and GX series balances
  • 1 year warranty
More About this Item
This can output analog voltage in the following modes : "mode where the specified digits of the weighing value are converted to voltage" and "mode where weighing value is converted to voltage in range between gross zero or net zero and full scale". The voltage output range can be selected using the "0V /0.2V " switch on the panel. It can be selected "0 to 1V" range and "0.2V to 1V" range. The factory setting is "0 to 1V" range.
1 Year Warranty
Specifications
  • DescriptionAnalog Output Interface