A&D Weighing GXA-03 Additional RS-232C Interface
Representative image only.
A&D Weighing GXA-03 Additional RS-232C Interface
A&D Weighing | Mfr: GXA-03| Item: 1110958
Communicate with your A&D balance using compatible output and relay kits
Call For Price
Call For Price
Buy It Together
Current Item
A&D Weighing GXA-03 Additional RS-232C Interface
A&D Weighing GXA-03 Additional RS-232C Interface
1110958
Call For Price
+ =
Total Price 336000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Compatible with A&D Apollo GF and GX series balances
  • 1 year warranty
More About this Item
Commands from the PLC or other devices installed in the factory environment can be used to make the following operations : output measurement data from the balance, input parameters into the balance, control the balance, reference parameters of the balance, etc. The RS-232C is equipped with a data format to manage the balance in accordance with GLP.
1 Year Warranty
Specifications
  • DescriptionAdditional RS-232C Interface