A&D Weighing AX-43 Temperature Calibrator for Moisture Balances
Representative image only.
A&D Weighing AX-43 Temperature Calibrator for Moisture Balances
A&D Weighing | Mfr: AX-43| Item: 1110717
Easily adjust drying temperature of pan
₹164,000.00INR / Each
₹164,000.00INR / Each
Product Description
Key Features
  • 2 year warranty
More About this Item
Provides calibration of drying temperature for MX50 and MS70 moisture balances. The temperature calibrator adjusts the drying temperature on the pan. Put the sensor on the pan and input measurement data at 100°C and 160°C. Each adjustment needs 15 minutes. The buzzer sounds at the end.
Two year warranty
Specifications
  • DescriptionTemperature Calibrator for Moisture Balances