A&D Weighing AD-8920A Universal Remote Display
Representative image only.
A&D Weighing AD-8920A Universal Remote Display
A&D Weighing | Mfr: AD-8920A| Item: 1111480
Enhance the versatility of your A&D Weighing Balance
₹81,000.00INR / Each
₹81,000.00INR / Each
Product Description
Includes
AC adapter and .munication cable
Key Features
  • Recognizes the balance/scale baud setting automatically
  • Large and bright VFD display
  • 1 year warranty
More About this Item
Choose from a wide range of accessories compatible with your A&D Weighing Balance for optimal performance.
Connects to A&D’s balances and scales using either RS-232C or current loop.
1 Year WarrantyConformite Europeenne
Specifications
  • RS-232 InterfaceYes
  • DescriptionUniversal Remote Display