A&D Weighing AD-1689 Tweezers For Calibration Weight
Representative image only.
A&D Weighing AD-1689 Tweezers For Calibration Weight
A&D Weighing | Mfr: AD-1689| Item: 1110992
Enhance the versatility of your A&D Weighing Balance
₹57,000.00INR / Each
₹57,000.00INR / Each
Product Description
Key Features
  • Compatible with A&D GF and GX series balances
  • 1 year warranty
More About this Item
Choose from a wide range of accessories compatible with your A&D Weighing Balance for optimal performance.
These tweezers that are ideally suited for manipulating calibration weights (10 ~ 500g). With a length of 210mm, the tweezers allow a weight to be placed in a weighing pan without having to put a hand inside the
chamber. This prevents convection flow that is normally attributed to heat coming from the hand. Each tweezer is made from austenitic stainless steel (SUS304), therefore heat from the hand is unlikely to be transmitted to
the weight via the tweezers. This is especially effective for the precise measurements required of analytical balances.
1 Year Warranty
Specifications
  • DescriptionTweezers For Calibration Weight