A&D Weighing AD-1688 External Data Logger for Balances; 5000 Reading Memory
Representative image only.
A&D Weighing AD-1688 External Data Logger for Balances; 5000 Reading Memory
A&D Weighing | Mfr: AD-1688| Item: 1111047
₹93,000.00INR / Each
₹93,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Data logger stores up to 5000 readings and exports to excel for analysis.
1 Year WarrantyConformite Europeenne
Specifications
  • Accessory TypeData Logger
  • MaterialPlastic
  • USB InterfaceYes
  • RS-232 InterfaceYes
  • Height (in)4 1/16
  • Width (in)2 1/8
  • Depth (in)5/8
  • DescriptionExternal Data Logger for Balances; 5000 Reading Memory