A&D Weighing AD-1654 Density Determination Kit for HR-AZ, HR-A, FZ- I, and, FX- i Series Balances
Representative image only.
A&D Weighing AD-1654 Density Determination Kit for HR-AZ, HR-A, FZ- I, and, FX- i Series Balances
A&D Weighing | Mfr: AD-1654| Item: 1111475
Use your balance to determine density with these convenient kits
₹251,000.00INR / Each
₹251,000.00INR / Each
Buy It Together
Current Item
A&D Weighing AD-1654 Density Determination Kit for HR-AZ, HR-A, FZ- I, and, FX- i Series Balances
A&D Weighing AD-1654 Density Determination Kit for HR-AZ, HR-A, FZ- I, and, FX- i Series Balances
1111475
₹251,000.00INR / Each
+
Cole-Parmer RS-232 to USB Cable
1010083
₹9,900.00INR / Each
=
Total Price 260900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Expand the capabilities of your balance with these convenient kits
  • Ensure the kit corresponds to your correct A&D Weighing balance
  • 3 month warranty
More About this Item
These density determination kits, when combined with one of the A&D Weighing electronic balances, allows the density of a solid and liquid to be measured easily. Balances sold separately.
3 Month WarrantyConformite Europeenne
Specifications
  • DescriptionDensity Determination Kit ,