7600 Open Door Sensor
Representative image only.
7600 Open Door Sensor
| Mfr: 7669TR| Item: 0750429
Open-door sensors prevent accidental temperature excursions
₹17,000.00INR / Each
₹17,000.00INR / Each
Product Description
Key Features
  • Detects whether a door is open or closed
  • 25 mm (1") gap distance
  • Includes a 9-3/4-ft long cable
More About this Item
Each data logger can support up to two open-door sensors. Sensors will alert if a door is accidentally left open.
Specifications
  • Compatible WithTraceable® 7600 Smart Wi-Fi Data Loggers
  • DescriptionDoor Sensor