3M Dual Reader Safety Glasses, 2.5x top and bottom diopters
Representative image only.
3M Dual Reader Safety Glasses, 2.5x top and bottom diopters
3M | Mfr: 11459-00000-20| Item: 8647435
Dual magnifiers on the top and bottom of the lens make close work easier
₹5,100.00INR / Each
₹5,100.00INR / Each
Product Description
Key Features
  • Adjustable temple lengths and soft nose bridge for comfortable, customized fit
  • ANSI Z87.1-2003 compliant
More About this Item
Ideal for workers that require reading glasses, but need eye protection as well. Dual magnifiers on the top and bottom of the lens make it easier when you have to look up and down at your work. Polycarbonate lens provides 99.9% UV protection.
Specifications
  • Frame ColorBlack/Silver
  • Lens ColorClear
  • ComplianceANSI Z87.1-2003
  • Magnification+2.5
  • DescriptionDual Reader Safety Glasses, 2.5x top and bottom diopters