3M 11380-00000-20 Eyewear, Silver/Black frame, Clear lens
Representative image only.
3M 11380-00000-20 Eyewear, Silver/Black frame, Clear lens
3M | Mfr: 3MR11380-00000| Item: 8647260
Three lens angles for a custom fit
₹2,100.00INR / Each
₹2,100.00INR / Each
Product Description
Key Features
  • Soft temple tips provide comfort for long periods of wear
  • Soft, universal-fit nosebridge
More About this Item
High-impact polycarbonate lens blocks 99.9% UV and features antifog coating. Temple is adjustable to three lengths. Compliance: ANSI Z87.1-2003, CSA Z94.3
Specifications
  • Frame ColorBlack/Silver
  • Lens ColorClear
  • ComplianceANSI Z87.1-2003; CSA Z94.3
  • DescriptionEyewear, Silver/Black frame, Clear lens