Tubing ID (mm)
Tubing OD (in)
Tubing OD (mm)
Tubing ID (in)
Min Temperature (° C)
Wall Thickness (in)
Wall Thickness (mm)
Viton
Viton
Viton tubing is a type of high-performance, fluoropolymer-based tubing that is known for its exceptional resistance to chemicals, heat, and weathering. Viton tubing is often used in industrial, laboratory, and other applications where durability and resistance to harsh conditions are required.