Readycult
Readycult
Readycult coliform snap pack is a rapid presence/absence test for the detection of total coliforms and E. coli in drinking water. This is an EPA-approved testing protocol. Dropper bottle is also available. We are a Readycult authorized distributor.
Sorry, we are having difficulties finding a match for the term(s) you have entered. Please clear your results and try again.