Versatile New Cole-Parmer StableTemp Thermal Cyclers Meet Research and Academic Lab Needs
10/5/2015
Nongradient thermal cycler uses a Peltier heater system to produce uniform temperature
Vernon Hills, Ill., October 5, 2015—The new Cole-Parmer® StableTemp™ Thermal Cyclers enable scientists to program PCR processes for easier operation. The units include two- and three-step PCR, RT-PCR, chill, and ligation program templates. Incremental and decremental temperatures and step hold times increase programming versatility.
For greater flexibility, users can switch out thermal blocks to adapt to their changing lab needs. The units’ heated lid features an etched foil heater with adjustable temperature and pressure to protect samples. The rotating dial, which lowers the silicone membrane over sample tubes, adjusts pressure.
Safety features include password protection of saved setting configurations and temperature cycle programs to prevent tampering and accidents. The thermal cyclers are designed to be workhorses in busy environments—multiple units can be daisy-chained together when used with user-supplied control software. Blocks are interchangeable for a quick change in testing protocol.
Personal/Compact Models have a small footprint to conserve bench space or fume hood use. These cyclers are designed for smaller sample batches, and feature faster heating and cooling rates than the full-size models. Full-Size Models are best suited for high throughput applications.
View the Cole-Parmer® StableTemp™ Thermal Cyclers at ColeParmer.com/21497. For more information, call 800-323-4340.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, Digi-Sense™, Environmental Express®, Ismatec®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL, 60061. In the US, call 1-800-323-4340. International customers, call 1-847-549-7600. Visit us at www.coleparmer.com.
Download 300 DPI 93943_21_27_angle.jpg Image