New Masterflex® Solve-Flex® tubing from Cole-Parmer revolutionises where you can use peristaltic pumps
Peristaltic pumps are the fastest growing positive displacement pump technology. Simple setup and maintenance, coupled with accurate dosing, have made them a preferred choice in the pharma and food markets.
Adoption of peristaltic pumps within the chemical industry has accelerated as advances in tubing design offer longer maintenance free periods. New tubing formulation also allow transfer and dosing of harsh chemicals not previously compatible with peristaltic pump tubing.
Cole-Parmer’s innovative new flexible tubing offers the chemical inertness of a fluoropolymer. Masterflex Solve-Flex tubing has a fluoropolymer-based liner which is plasticizer-free, hydrophobic, and resists the absorption of aqueous fluids. It also offers low spallation and assured purity of fluid. A flexible outer jacket extends the pumping life of the tubing.
Masterflex Solve-Flex Pump Tubing can be used for corrosive fluids, solvents and aggressive chemicals, including organic solvent lines, ink and solvent production, diagnostic testing, specialty chemical processing, and ink and toner feed lines. It also provides an option for transferring essential oils in the food processing and pharmaceutical industry. The flexible tubing solves the challenge of hard-to-match peristaltic pump applications.
Masterflex Solve-Flex Pump Tubing meets USP Class VI requirements. It is available in long continuous lengths, allowing for an unobstructed fluid path. For more information, call 020-8574-7556 or go to Download 300 DPI 96446_17.1.jpg Image