Vernon Hills, Ill.— Ensure effective cold chain management with the one button plug-and-play Digi-Sense TraceableOne Single-Use USB Temperature Data Logger with Calibration from Cole-Parmer. Each single-use data logger is traceable to NIST standards and comes with a Traceable® certificate which ensures accuracy. The data loggers are ideal for anyone who needs to make sure the temperature or temperature and humidity of their items remain at set parameters during transport, storage or experiments in the food, pharmaceutical, or laboratory industries.
These compact, data loggers fit anywhere—even inside a small cooler and include many other useful features. They are pre-programmed to measure typical temperature ranges of interest with alarm set-points that allow a quick check for outliers. The data loggers auto generate PDFs when plugged in to a computer. No software or driver is required for set-up or download data. They are low cost and easy to use. Flashing red and green LEDs indicate logger is recording data, stopped recording, and if an alarm has been triggered. Logging duration runs from either 10, 30, 60, or even 90 days. Logging intervals update either every 1, 3, 6, or 10 minutes. A waterproof design (IP67) eliminates concerns when shipping materials with cold packs, ice, or in damp environments.
For more information on Digi-Sense TraceableOne Single-Use Data Loggers with Calibration, go to ColeParmer.com or call our technical experts at 1-800-323-4340.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Ismatec®, Oakton®, Digi-Sense™, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 1-800-323-4340. International customers, call 1-847-549-7600. You can also visit www.coleparmer.com.
Popular Categories
- Balances and Scales
- Analytical Balances
- Bench and Floor Scale Bases
- Bench and Floor Scales
- Counting Scales
- Crane and Hanging Scales
- Health Scales
- Indicators for Scales
- Mechanical Balances
- Moisture Balances
- Platform and Pallet Scales
- Production Weighing
- Semi and Micro Analytical Balances
- Toploading Balances
- Weighing Accessories
- Weighing Dishes
- Calibration
- Electrical Products
- Batteries and Battery Testers
- Cable Testers
- Cable Ties
- Clamp Meters
- Electrical Accessories
- Electronics Enclosures
- Function Generators
- Gauss Meters
- Insulation Testers
- LCR Meters
- Megohmmeters
- Motor Rotation Testers
- Multimeters
- Oscilloscopes
- Outlet Strips
- Power Meters and Analyzers
- Power Supplies
- Resistance Testers
- RF Signal Generators
- Static Control Products
- Surge Protectors
- Test Leads and Probes
- Voltage Detectors
- Filtration Products
- Fluid Handling
- Furniture
- Heating and Cooling Equipment
- Laboratory Equipment
- Baths and Circulators
- Burettes (Burets)
- Calorimeters
- Centrifuges
- Chromatography Products
- Desiccators
- Digestion Systems
- Dispensers
- Distillation
- Electrophoresis Products
- Electroporation Products
- Environmental Samplers
- Extraction Apparatus
- Fermentation Products
- Flame Photometers
- Fluid Bed Dryers
- Freeze Dryers
- Glassware Washers
- Histology Products
- Homogenizers and Ultrasonic Processors
- Humidifiers and Dehumidifiers
- Lab Shakers
- Laboratory Evaporators
- Laboratory Sieves and Sieve Shakers
- Leachate Equipment
- Lighting and Illumination
- Magnetic Stirrers and Hot Plates
- Microscopes and Magnification
- Mixers and Blenders
- Particle Counters
- Pipettes and Pipettors
- Refractometers
- Thermal Cyclers
- Titrators
- Ultrasonic Cleaners
- Vacuums
- Water Purification Products
- Laboratory Supplies
- Air Sampling
- Animal Research Products
- Bags
- Beakers
- Books and Reference Materials
- Bottles and Jars
- Calculators
- Carboys
- Chemistry Kits
- Clean Room Stationery
- Containers
- Cuvettes
- Dialysis Products
- Dippers
- Distillation Supplies
- Field Sampling
- Flasks
- Graduated Cylinders
- Ice Buckets and Ice Pans
- Lab Cleaning Products
- Lab Organizers
- Labels and Label Printers
- Laboratory Dishes
- Laboratory Timers and Clocks
- Labware
- Microbiology Products
- Microplate Products
- Mortars and Pestles
- Parts and Accessories
- Racks
- Sampling Wipes
- Shippers
- Syringes
- Tubes
- Vials
- Watch Glasses
- Material Handling and Storage
- Material Testing and Physical Testing
- Pressure and Vacuum Instruments
- Process Controllers
- Pumps
- Safety Products
- Air Quality Monitors
- Arc Flash Products
- Combustion Analyzers
- Eyewash
- Eyewear and Safety Glasses
- Floor Mats and Matting
- Gas Detectors
- Gloves
- Hearing Protection
- Lockout and Tagout Product
- Personal Monitor Badges
- Radiation Detectors
- Refrigerant Products
- Respirators and Masks
- Safety Apparel
- Safety Shields
- Safety Signs and Labels
- Solvent Safety
- Sound Meters
- Spill Control
- Temperature Measurement Products
- Test and Measurement Products
- Water Quality Products
- Colorimeters
- Colorimetry Reagents
- Conductivity, Resistivity, and TDS Meters
- Dissolved Oxygen Meters
- Dye Tracers
- Electrodes
- Flocculation Testers
- Multiparameter Meters
- pH Buffers
- pH Meters
- Turbidity Accessories
- Turbidity Meters
- Water Quality Accessories
- Water Quality Standards, and Solutions
- Water Quality Test Strips