TerraSpray® Rinse Station from Cole-Parmer - alternative to expensive automated systems or manual cleaning
7/25/2012
Compact design fits in small spaces or on laboratory benchtops and keeps water contained—eliminating wet floors and slippery hazards from laboratories and workshops
The TerraSpray® Rinse Station from Cole-Parmer is a cost-effective choice for cleaning all styles of vessels and carboys up to 200 litres. The station connects directly to water inlet and drain—no electricity needed.
Select from countertop or floor models. The countertop model cleans vessels up to 38.1 cm (15”) in diameter. Operate by the hand-controlled valve on the side of the unit. The floor model cleans vessels up to 48.3 cm (19”) in diameter. A foot-controlled valve on the bottom platform turns rinse on and off. Both models are easy to use with quick set-up.
The units are crafted of a roto-molded polyethelyne top panel and fabricated polyethelyne base and legs. Valves are made of polypropylene with Viton™ elastomers and a Kynar™ coated stainless steel spring. Spray head is manufactured with PVDF.
Further information available on online. To discuss your specific applications and requirements call 0500-345-300 and ask to speak to an Application Specialist.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support.
For more information, contact Cole-Parmer, Unit 3 River Brent Business Park, Trumpers Way, Hanwell, London W7 2QA. Call +44-(0)20-8574-7556.
Download 300 DPI 98965_08_app.jpg Image