Safe, convenient option for the reporting of cyanide
Charleston, S.C. June 3, 2015—Small-volume distillations are easy with the new SimpleDist® Micro from Environmental Express®. Smaller sample sizes save time—the unit distills 26 samples simultaneously. It also eliminates the need for costly glassware through the use of disposable polypropylene SimpleDist tubes. No cooling water is required.
With proven HotBlock® dependability, the SimpleDist Micro heating block is made from PTFE-coated graphite enclosed in a durable Kydex® housing with a built-in controller.
The EPA has approved the use of micro distillation systems with disposable tubes for reporting on cyanide. The system can also adapt to report on ammonia and phenols as well.
For more information, go to www.ColeParmer.com/21431 or call 800-343-5319.
Environmental Express is a leading developer, manufacturer and distributor of environmental laboratory equipment and consumable supplies for commercial, governmental, industrial and academic laboratories worldwide. The company provides an entire range of laboratory products used in applications such as water/wastewater analysis, oil and grease analysis, metals analysis and hazardous waste analysis. We pride ourselves on providing innovative products, superior technical support, knowledgeable customer service and same day shipping.
The company’s products are specified in over 30 EPA standards methods and range from equipment including the award-winning AutoBlock® System for Metals Digestions and SimpleDist® System for Ammonia and Cyanide Distillations to disposable coated BOD bottles and preweighed ProWeigh® filters.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, Digi-Sense™, Environmental Express® and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 800-323-4340. International customers, call 847-549-7600. Visit us at www.ColeParmer.com.
Download 300 DPI ee-pittonc-simpledist_logo.jpg Image