New easy setup and use Digital Gear Pump Drive by Cole-Parmer
With a larger, brighter interface, analog remote control, and simple programming, the new Cole-Parmer Digital Gear Pump Drive for Micropump A-Mount Pump Head is easy to operate for all automated process applications.
Hands-free dispensing is easy, simply set time delay between cycles. The batch count function counts down and displays batches completed while the cumulative volume or totaliser tracks amount dispensed across multiple cycles. The four operating modes—continuous run, timed dispense, copy dispense, and volume dispense—can be monitored on the graphical, multilanguage LCD display, for continual performance monitoring.
A pulse-free flow is delivered when used with the Micropump A-mount pump head. Its turn down ratio is 100:1, while the flow range is from 0.612 to 4212 mL/min. The metering pump drive is IP33-rated to ensure splash- and dust- resistance. Smooth contours allow for easy cleaning. Stack up to three drives to conserve bench-top space.
To view the Cole-Parmer Digital Gear Pump Drive for Micropump A-Mount Pump Head, go to ColeParmer.co.uk/21335 or, for more information, email [email protected] or call 020-8574-7556
Download 300 DPI 75211_70_left.jpg Image