Portable secure cold storage for medical and clinical media in extreme conditions
1/8/2013
New Shuttle Portable Ultra-Low Temperature Freezers from Cole-Parmer
Now, a lightweight portable freezer with ultra-low -86 C to -20 C temperature range can go places a conventional compressor freezer cannot go. The new energy-efficient Shuttle Portable Ultra-Low Temperature Freezer from Cole-Parmer offers secure mobile transport of high-value vaccines or biological specimens. Its unique functionality is ideal for biorepositories, medical/surgical suites, pathology laboratories, tissue banks, regenerative medicine, and other applications in clinical or diagnostic medicine, research, and pharmaceutical distribution.
A Stirling engine operates the low temperature freezer, which requires no compressors, dry ice, or LN2, and expends the same volume of energy as a conventional light bulb. The Humm™ free-piston Stirling engine uses two moving parts that float on helium gas bearings to eliminate contact wear. The linear driver modulates engine power to deliver cooling on demand without on/off cycling. The thermosiphon evaporator freezes contents. A microcharge of environmentally friendly cooling medium creates a uniform ultra-low temperature via a continuous process of evaporation and condensation within the sealed tube.
Weighing 42 lbs, the portable freezer is efficient for both benchtop and field uses. A weather-resistant touchpad offers digital temperature display indicating interior temperature and error codes. It includes temperature presets of -20 C;, -40 C, or -86 C and a system lock indicator. Adjust by 1 C intervals. Features visual and audible alarms.
The freezer’s rugged cabinet withstands travel. Its composite vacuum-insulation panels with CFC-free foam protect contents from high ambient temperatures. When not in use, the cabinet can be stored on side, end, or upside down. A double gasket seals the interior chamber and provides dual protection against moisture and heat gain. Molded-in handles are easy to grip. Operates on both AC/DC (12V) power by simply switching the included power cord. For more information, call 0500-345-300 or visit ColeParmer.co.uk/19664.
Download 300 DPI 44199_05.jpg Image