Independently control flow and direction of each channel in a single compact unit
Vernon Hills, Ill., February 4, 2014—Experience maximum pumping flexibility with the Ismatec® ICC 4-Channel 8-Roller Peristaltic Pump from Cole-Parmer. This four-channel peristaltic pump lets users individually control the flow of each channel from the keypad or via computer. Each channel also offers bidirectional flow for even more versatility.
The micropump delivers continuous pumping or precision dispensing up to 35 mL/min. Independent channel calibration minimizes tube-to-tube differences, resulting in high calibration accuracy. Easy-to-use tubing cassettes enable fast changeovers.
The pump contains a USB interface for quick connections. To view the Ismatec® 4-Channel Independent Channel Control Peristaltic Pump, go to ColeParmer.com/20788 or call 800-323-4340 for more information.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 800-323-4340. International customers, call 847-549-7600. Visit us at www.ColeParmer.com
Download 300 DPI 78001_80.jpg Image