Buffalo, NY, August 29, 2022 — The recently launched ZeptoMetrix® NATtrol™ external run control for Flu/RSV/SARS-CoV-2: NATFRC-ERC is now available for order and any pre-orders will be filled and shipped. This third-party molecular quality control can be used by laboratories to monitor the continued performance of their multiplexed respiratory assays. With target concentrations of 15,000 cp/mL for all four analytes, the launch of NATFRC-ERC represents an exciting addition to the ZeptoMetrix respiratory product line.
“The flu season is here. It’s our top priority to make sure laboratories are equipped with the materials needed to accurately detect and differentiate the most common respiratory infections,” said Evangeline Gonzalez, Vice President/General Manager of ZeptoMetrix.
Adding this external run control to the ZeptoMetrix portfolio expands a line of easy-to-use, refrigerated, inactivated whole organism controls designed to quickly and safely assure that testing instruments are providing accurate results.
As laboratories continue to adapt to meet the challenges of the COVID-19 pandemic, ZeptoMetrix is assisting with its broad portfolio of quality control materials including not only respiratory controls but also Women’s Health, Gastrointestinal, and Healthcare-Associated Infections.
Get more information on the respiratory controls and associated products at www.zeptometrix.com.
Popular Categories
- Balances and Scales
- Analytical Balances
- Bench and Floor Scale Bases
- Bench and Floor Scales
- Counting Scales
- Crane and Hanging Scales
- Health Scales
- Indicators for Scales
- Mechanical Balances
- Moisture Balances
- Platform and Pallet Scales
- Production Weighing
- Semi and Micro Analytical Balances
- Toploading Balances
- Weighing Accessories
- Weighing Dishes
- Calibration
- Electrical Products
- Batteries and Battery Testers
- Cable Testers
- Cable Ties
- Clamp Meters
- Electrical Accessories
- Electronics Enclosures
- Function Generators
- Gauss Meters
- Insulation Testers
- LCR Meters
- Megohmmeters
- Motor Rotation Testers
- Multimeters
- Oscilloscopes
- Outlet Strips
- Power Meters and Analyzers
- Power Supplies
- Resistance Testers
- RF Signal Generators
- Static Control Products
- Surge Protectors
- Test Leads and Probes
- Voltage Detectors
- Filtration Products
- Fluid Handling
- Furniture
- Heating and Cooling Equipment
- Laboratory Equipment
- Baths and Circulators
- Burettes (Burets)
- Calorimeters
- Centrifuges
- Chromatography Products
- Desiccators
- Digestion Systems
- Dispensers
- Distillation
- Electrophoresis Products
- Electroporation Products
- Environmental Samplers
- Extraction Apparatus
- Fermentation Products
- Flame Photometers
- Fluid Bed Dryers
- Freeze Dryers
- Glassware Washers
- Histology Products
- Homogenizers and Ultrasonic Processors
- Humidifiers and Dehumidifiers
- Lab Shakers
- Laboratory Evaporators
- Laboratory Sieves and Sieve Shakers
- Leachate Equipment
- Lighting and Illumination
- Magnetic Stirrers and Hot Plates
- Microscopes and Magnification
- Mixers and Blenders
- Particle Counters
- Pipettes and Pipettors
- Refractometers
- Thermal Cyclers
- Titrators
- Ultrasonic Cleaners
- Vacuums
- Water Purification Products
- Laboratory Supplies
- Air Sampling
- Animal Research Products
- Bags
- Beakers
- Books and Reference Materials
- Bottles and Jars
- Calculators
- Carboys
- Chemistry Kits
- Clean Room Stationery
- Containers
- Cuvettes
- Dialysis Products
- Dippers
- Distillation Supplies
- Field Sampling
- Flasks
- Graduated Cylinders
- Ice Buckets and Ice Pans
- Lab Cleaning Products
- Lab Organizers
- Labels and Label Printers
- Laboratory Dishes
- Laboratory Timers and Clocks
- Labware
- Microbiology Products
- Microplate Products
- Mortars and Pestles
- Parts and Accessories
- Racks
- Sampling Wipes
- Shippers
- Syringes
- Tubes
- Vials
- Watch Glasses
- Material Handling and Storage
- Material Testing and Physical Testing
- Pressure and Vacuum Instruments
- Process Controllers
- Pumps
- Safety Products
- Air Quality Monitors
- Arc Flash Products
- Combustion Analyzers
- Eyewash
- Eyewear and Safety Glasses
- Floor Mats and Matting
- Gas Detectors
- Gloves
- Hearing Protection
- Lockout and Tagout Product
- Personal Monitor Badges
- Radiation Detectors
- Refrigerant Products
- Respirators and Masks
- Safety Apparel
- Safety Shields
- Safety Signs and Labels
- Solvent Safety
- Sound Meters
- Spill Control
- Temperature Measurement Products
- Test and Measurement Products
- Water Quality Products
- Colorimeters
- Colorimetry Reagents
- Conductivity, Resistivity, and TDS Meters
- Dissolved Oxygen Meters
- Dye Tracers
- Electrodes
- Flocculation Testers
- Multiparameter Meters
- pH Buffers
- pH Meters
- Turbidity Accessories
- Turbidity Meters
- Water Quality Accessories
- Water Quality Standards, and Solutions
- Water Quality Test Strips



