New Vortex-Genie® 2 Multitube Holders Ideal for High-Speed Mixing
2/9/2013
Cole-Parmer offers three new high-speed multitube holders as accessories for the original laboratory standard Vortex-Genie® 2 and Vortex-Genie® Pulse Mixers
The new tube holders are optimised to achieve the highest vortex action when mixing only a small number of tubes. The Vortex-Genie® 2 Multi-Tube Holders are available for tube sizes 50mL, 15mL, or microtubes. The tubes are securely fastened and achieve true vortex action in each tube.
With many years of proven performance in laboratories all over the world, the Vortex-Genie®2 Shaker boasts a broad range of attachments of various shapes, sizes, and materials allowing for mixing of almost all common tubes or containers for hands-free or touch operation.
Its variable-speed control allows slow-speed shaking up to high-speed vortex action. In addition, each attachment is designed to produce different mixing actions, allowing users to choose the correct action for each application. Tough metal housing, combined with "just enough weight", provides a stable platform for all types of mixing. For more information visit ColeParmer.co.uk/19791 or contact a Cole-Parmer Applications Specialist at 0500-345-300 to discuss your specific requirements.
Download 300 DPI 04724_64.jpg Image