New Non-DEHP Lab Tubing from Cole-Parmer
9/25/2012
Nontoxic formulation is safe for contamination-free applications
The new Cole-Parmer® Non-DEHP Lab Tubing offers a versatile option for analytical instruments, food processing, and general laboratory uses. The tubing transfers liquids, gases, and inorganic chemicals found in lab, paint, pigment, and soap dispensing applications. Because it contains no DEHP (di(2-ethylhexyl)phthalate), it is free from leachates that may compromise the transferred material.
Made of clear PVC material, this tubing can be visibly inspected for flow monitoring. It is flexible, with a small bend radius, and installs easily on barbed fittings. This autoclavable tubing meets FDA CFR 21, Part 175.300 certification and is Proposition 65- and REACH-compliant. Use the Cole-Parmer Non-DEHP Lab Tubing for temperatures ranging from -58 to 165°F (-50 to 73°C). For more information, call 0500-345-300 or visit ColeParmer.co.uk/19498.
Download 300 DPI 96605_31.jpg Image