Optimize stirring needs and work space with Cole-Parmer® elements™ Modular Magnetic Stirrer
Vernon Hill, Ill. — The customizable elements modular magnetic stirrer from Cole-Parmer, can easily be configured to fit a work space and meet stirring needs—use from one up to 10 stirring plates at a time with one control unit. Attaching or removing a stir plate is easy by using a direct USB connection or with included cables. This stirrer is ideal for anyone who needs more stirring equipment but has minimal work space or doesn’t want a bunch of bulky stirrers sitting around in their laboratory.
The innovative stirrer hosts many useful features. The control unit has a touch and slide bar to easily operate the parameters. Real-time parameters can be viewed on the LCD screen. Multiple stirring options allow a choice between one direction or reversing every 45 seconds for thorough mixing. A built-in timer can be set for up to nine hours of continuous operation. The speed, reverse mode and timing settings will be saved automatically when the instrument is turned off and will automatically be restored the next time the user turns on the instrument. Each stir plate has lights along the edge of the plate to accurately position a vessel and a corrosion resistant glass stir top. Both the control unit and the stirrer unit have 3 sockets that can be connected to another stirrer unit or power line.