New Method in Sample Preparation with PBI Shredder SG3™ System available now from Cole-Parmer
11/27/2012
Unique device uses a closed system ideal for molecular and cellular biological analysis
Better sample integrity and longer DNA is now available with the new PBI Shredder SG3 from Cole-Parmer. Ideal for the extraction of DNA, RNA, protein, mitochondria, and small molecules, the device creates a closed system in which to safely prepare samples. It is also portable for easy and efficient field collection.
Its closed system operates by using pressure, which forces the sample against the lysis plate in the tubes for low-shear cell disruption. The resulting samples exceed the yields of those derived from traditional labor-intensive methods such as manual mortar and pestle preparation. The samples also offer better integrity than bead beating methods. Higher reproducibility and lower cost in sample preparation make this efficient new device a standout option for research labs, forensics, cryogenics, medical diagnostics, and more. It also offers full chain-of-custody tracking from collection to testing.
The PBI Shredder SG3 System is easy to use and compatible with lab buffers and reagents already present on most labs. Its heavy-duty driver features long-lasting rechargeable lithium batteries for uninterrupted operation. A three-position setting level enables users to create a standard for reproducible results among various operators. Unique polypropylene shredder tubes create a closed system for sample collection, preparation, storage, and transporting. This design prevents sample cross-contamination and ensures safety in processing hazardous materials. Tubes are also available with a metal lysis disk for harder sampler processing, such as seeds or ticks.
For more information on the PBI Shredder SG3 System for sample prep and analytical testing, call 0500-345-300 or visit ColeParmer.co.uk/19586.
Download 300 DPI 04749_00.jpg Image