Jenway Launches Genova Bio Spectrophotometer for Life Science Applications
Scanning diode array technology measures wavelength range simultaneously in under 3 seconds
September 6, 2017, Vernon Hills, Ill. – The new Jenway Genova Bio UV/visible spectrophotometer, from Cole-Parmer, has been designed for fast and easy use in life science applications. This compact and lightweight spectrophotometer is ideal for molecular biology laboratories, biotechnology, biochemistry and cell biology applications. Laboratory managers worldwide now have the ability to equip their labs with a state-of-the-art, reliable spectrophotometer at an affordable price.
This spectrophotometer leverages scanning diode array technology, which uses electrical scanning to measure the wavelength range (198 nm to 800 nm) simultaneously in under three seconds. Ideal where multiple wavelengths are required for purity ratios. The instrument features a small footprint and large intuitive color touch-screen interface for simple operation in the lab. It is compatible with a wide range of small volume cuvettes, making it an excellent tool for measuring the purity and concentration of DNA, RNA and other biological samples. To optimize sample preparation, the Genova Bio is preprogrammed with methods for the quantification of nucleic acids and proteins. The instrument also has a preprogrammed method for measuring optical density of bacterial cell cultures such as E. coli and yeast cells, enabling scientists to measure cell growth before cell harvesting.
The Genova Bio replaces the Genova Plus and completes the Jenway 72 Series spectrophotometers in Jenway’s comprehensive range of spectrophotometers that features the 7200, 7205 and the Genova Bio spectrophotometers.
For more information on the Genova Bio spectrophotometer, visit ColeParmer.com
Cole-Parmer has been a leading global manufacturer and distributor of fluid handling products, instrumentation, equipment, and supplies since 1955. Our portfolio including popular brand names such as Masterflex®, Ismatec®, Oakton®, Digi-Sense™, Environmental Express® and more, are sold around the globe through direct sales, multichannel marketing outlets, and a strong network of international dealers. Cole-Parmer also features an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 1-800-323-4340. International customers, call 1-847-549-7600.
Popular Categories
- Balances and Scales
- Analytical Balances
- Bench and Floor Scale Bases
- Bench and Floor Scales
- Counting Scales
- Crane and Hanging Scales
- Health Scales
- Indicators for Scales
- Mechanical Balances
- Moisture Balances
- Platform and Pallet Scales
- Production Weighing
- Semi and Micro Analytical Balances
- Toploading Balances
- Weighing Accessories
- Weighing Dishes
- Calibration
- Electrical Products
- Batteries and Battery Testers
- Cable Testers
- Cable Ties
- Clamp Meters
- Electrical Accessories
- Electronics Enclosures
- Function Generators
- Gauss Meters
- Insulation Testers
- LCR Meters
- Megohmmeters
- Motor Rotation Testers
- Multimeters
- Oscilloscopes
- Outlet Strips
- Power Meters and Analyzers
- Power Supplies
- Resistance Testers
- RF Signal Generators
- Static Control Products
- Surge Protectors
- Test Leads and Probes
- Voltage Detectors
- Filtration Products
- Fluid Handling
- Furniture
- Heating and Cooling Equipment
- Laboratory Equipment
- Baths and Circulators
- Burettes (Burets)
- Calorimeters
- Centrifuges
- Chromatography Products
- Desiccators
- Digestion Systems
- Dispensers
- Distillation
- Electrophoresis Products
- Electroporation Products
- Environmental Samplers
- Extraction Apparatus
- Fermentation Products
- Flame Photometers
- Fluid Bed Dryers
- Freeze Dryers
- Glassware Washers
- Histology Products
- Homogenizers and Ultrasonic Processors
- Humidifiers and Dehumidifiers
- Lab Shakers
- Laboratory Evaporators
- Laboratory Sieves and Sieve Shakers
- Leachate Equipment
- Lighting and Illumination
- Magnetic Stirrers and Hot Plates
- Microscopes and Magnification
- Mixers and Blenders
- Particle Counters
- Pipettes and Pipettors
- Refractometers
- Thermal Cyclers
- Titrators
- Ultrasonic Cleaners
- Vacuums
- Water Purification Products
- Laboratory Supplies
- Air Sampling
- Animal Research Products
- Bags
- Beakers
- Books and Reference Materials
- Bottles and Jars
- Calculators
- Carboys
- Chemistry Kits
- Clean Room Stationery
- Containers
- Cuvettes
- Dialysis Products
- Dippers
- Distillation Supplies
- Field Sampling
- Flasks
- Graduated Cylinders
- Ice Buckets and Ice Pans
- Lab Cleaning Products
- Lab Organizers
- Labels and Label Printers
- Laboratory Dishes
- Laboratory Timers and Clocks
- Labware
- Microbiology Products
- Microplate Products
- Mortars and Pestles
- Parts and Accessories
- Racks
- Sampling Wipes
- Shippers
- Syringes
- Tubes
- Vials
- Watch Glasses
- Material Handling and Storage
- Material Testing and Physical Testing
- Pressure and Vacuum Instruments
- Process Controllers
- Pumps
- Safety Products
- Air Quality Monitors
- Arc Flash Products
- Combustion Analyzers
- Eyewash
- Eyewear and Safety Glasses
- Floor Mats and Matting
- Gas Detectors
- Gloves
- Hearing Protection
- Lockout and Tagout Product
- Personal Monitor Badges
- Radiation Detectors
- Refrigerant Products
- Respirators and Masks
- Safety Apparel
- Safety Shields
- Safety Signs and Labels
- Solvent Safety
- Sound Meters
- Spill Control
- Temperature Measurement Products
- Test and Measurement Products
- Water Quality Products
- Colorimeters
- Colorimetry Reagents
- Conductivity, Resistivity, and TDS Meters
- Dissolved Oxygen Meters
- Dye Tracers
- Electrodes
- Flocculation Testers
- Multiparameter Meters
- pH Buffers
- pH Meters
- Turbidity Accessories
- Turbidity Meters
- Water Quality Accessories
- Water Quality Standards, and Solutions
- Water Quality Test Strips