Monitor vaccines and other important samples in transit via Bluetooth® technology and TraceableLive™
Vernon Hill, Ill. — Safely transport valuable materials such as reagents, clinical samples, blood, vaccines, tissues, pharmaceuticals and food to any location with the new Digi-Sense TraceableGo Data Logger now featuring a choice of three external probes. The new probes are designed to fit securely inside the carton while the data logger secures to the outside of the container for easy monitoring and access. There is now no need to open the container to read the data logger. These compact, durable data loggers fit almost anywhere—even inside a small cooler.
Three new probes are available for the data loggers to meet unique monitoring needs:
- External Bottle Probe is a temperature-buffered probe ideal for monitoring vaccines or other important samples in transit.
- External Bullet Probe is designed to monitor air and liquids, the sensor and cable may be completely immersed into the material.
- External RTD Probe monitors temperatures below –80°C ensuring critical samples are not compromised. It has a 3-m FEP-coated cable and 316 SS tip for rugged durability. Platinum RTD sensor provides a wider range and greater accuracy.
These handy data loggers with external probes are easy to operate. The probe simply plugs into the side of the data logger. The data loggers can be easily configurated from any Bluetooth-enabled device using the free TraceableGO app to set parameters and alarms and are capable of storing up to 90 events. ILAC MRA, NIST traceable calibration is included to ensure accurate, reliable measurements. Once the shipment is delivered, enable Bluetooth to export data to a smartphone, tablet, or computer and immediately see if shipment contents were exposed to conditions outside the set parameters.
Data logger owners can also subscribe to cloud-based TraceableLive service to upload data to the Cloud—ideal for tracking data from multiple loggers or to comply with regulatory agency requirements including 21 CFR Part 11 and EU Annex 11 compliance. This low-cost subscription provides unlimited trip data to be saved in the cloud and the ability to send a report to an email.
For more information on Digi-Sense Temperature and Temperature/Humidity Data Loggers with TraceableGO Bluetooth capability and external probes, go to coleparmer.com/Digi-Sense