Cole-Parmer Releases Updated Symmetry® EC Toploading Balances
With an updated design, the popular Cole-Parmer® Symmetry® EC Toploading Balances from Cole-Parmer are an even more exceptional choice in weighing. For general-purpose weighing applications, these laboratory balances or weigh scales offer accuracy and traceability, including a print function for GLP compliance. The balances now include a bigger screen and easier-to-use keypad. A no-hassle single-unit draft shield is more durable and the balance’s adjustable feet offer greater stability.
For operational ease, the balances feature a capacity tracker that warns of approaching overload conditions and a security lock-down port. The below-balance weigh hook can be used for density determination applications. An internal rechargeable battery offers convenient portability. The RS-232 interface allows for communication with a printer.
For more information on the Cole-Parmer® Symmetry® EC Toploading Balances, call 0500-345-300 or visit ColeParmer.co.uk/19663.
Download 300 DPI 20000_32.jpg Image