New Cole-Parmer® Syringe Pumps with Touch-Screens Create and Recall Configurations
3/19/2013
Three new models offer ultralow- to medium-level flow rates
Vernon Hills, Ill., March 19, 2013—For lab users seeking a convenient touch-screen interface, Cole-Parmer introduces three new syringe pumps. Each model uses touch-screen technology to quickly recall previous configurations or create new ones.
The Cole-Parmer® Touch-Screen Entry-Level Syringe Pumps are cost effective units that accommodate up to two syringes. They offer high precision fluid dosing with accuracy of ±0.5% and reproducibility of ±0.5%. View all operating parameters on their full-color touch screen.
The Cole-Parmer® Touch-Screen Continuous-Cycle Four-Syringe Pumps hold up to four syringes to cycle continuously back and forth in a push-pull action. As two syringes are infusing, two syringes are simultaneously withdrawing at the same rate. At the end of the set volume, the direction is automatically reversed and the next cycle begins. Offer accuracy of ±0.35% and reproducibility of ±0.5%. Daisy-chain up to 99 pumps together using the built-in RS-485.
The Cole-Parmer® Touch-Screen Nanoliter Infusion Syringe Pumps, are ideal for stereotaxic, micro manipulator, and other clamping devices. Use remote injector up to six feet away from the controller. Accurate, pulse-free flow from 3.66 pL/min to 3.818 mL/min; accuracy of ±0.5% and reproducibility of ±0.5%.
To find out more about any of these syringe pumps, call 800-323-4340 or visit < a href="http://www.coleparmer.com/Category/ColeParmer_Touch_Screen_Syringe_Pumps/58664?referred_id=19845">ColeParmer.com.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 800-323-4340. International customers, call 847-549-7600. Visit us at ColeParmer.com
Download 300 DPI 74905_34_R.jpg Image